Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MYH2 Polyclonal Antibody
Rabbit Polyclonal Antibody
$178.40 - $482.00
Specifications
Antigen | MYH2 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119293
|
Invitrogen™
PA5119293 |
200 μL |
Each for $482.00
|
|
|||||
PIPA5121752
|
Invitrogen™
PA5121752 |
20 μL |
Each for $178.40
|
|
|||||
Description
MYH2 Polyclonal Antibody for Western Blot, IHC (P)
MYH2 (Myosin-2) is associated with muscle contraction and is required for cytoskeleton organization.Specifications
MYH2 | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Mouse | |
MYH2 | |
2A myosin heavy chain; fast 2a myosin heavy chain; IBM3; inclusion body myopathy 3, autosomal dominant; MHC2A; Myh1; MYH2; MYH2A; MYHas8; MyHC-2A; MyHC-IIa; MYHSA2; Myhs-f; Myhsf1; Myhs-f1; myosin heavy chain 2; Myosin heavy chain 2a; myosin heavy chain IIa; myosin heavy chain type IIx; myosin heavy chain, skeletal muscle, adult 2; myosin, heavy chain 2, skeletal muscle, adult; myosin, heavy polypeptide 2, skeletal muscle, adult; myosin-2; MYPOP; skeletal muscle myosin heavy chain II-a; type IIA myosin heavy chain | |
MYH2 | |
Primary | |
Antigen Affinity Chromatography |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
0 | |
17882 | |
Recombinant protein Myosin Heavy Chain 2. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title