Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYH4 Antibody, Novus Biologicals™
SDP

Catalog No. NB259142 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB259142 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB259142 Supplier Novus Biologicals Supplier No. NBP323357
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MYH4 Polyclonal antibody specifically detects MYH4 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen MYH4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:2000
Formulation PBS, 1 mg/ml BSA and 50% glycerol
Gene Alias MyHC-2B, MyHC-IIb, myosin heavy chain 4, myosin heavy chain IIb, myosin heavy chain, skeletal muscle, fetal, myosin, heavy chain 4, skeletal muscle, myosin, heavy polypeptide 4, skeletal muscle, myosin-4
Host Species Rabbit
Immunogen A synthetic peptide (coupled to KLH) corresponding to amino acid residues at the C-terminus from mouse MYH4. This sequence has 100% identity with rat and human MYH4, and has significant homology to MYH1, MYH2, and MYH8. [UniProt# Q5SX39]
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4622
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.