Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYH6 Antibody (3-48) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB300284 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300284 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300284 Supplier Novus Biologicals Supplier No. NB300284

Mouse Monoclonal Antibody has been used in 9 publications

MYH6 Monoclonal specifically detects MYH6 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen MYH6
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 17593
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry 1:300, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:300
Gene Accession No. Q7Z406
Gene Alias alpha-MHC, ASD3, cardiomyopathy, CMD1EE, CMH14, Hypertrophic 1, MYHC, MYHCA, myHC-alpha, Myosin Heavy Chain 6, Myosin Heavy Chain, Cardiac Muscle Alpha Isoform, Myosin, Heavy Chain 6, Cardiac Muscle, Alpha, Myosin, Heavy Polypeptide 6, Cardiac Muscle, Alpha (Cardiomyopathy, Hypertrophic 1), myosin-6, SSS3
Gene Symbols MYH6
Host Species Mouse
Immunogen Full length native protein (purified) (Human).
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4624
Test Specificity This is myosin alpha and beta heavy chain specific. It binds to A-band of thick filament of cardiac and skeletal muscle myosin as determined by indirect immunofluorescence on cryocuts, and indirect immunoperoxidase staining of Carnoy fixed tissues.
Reconstitution Reconstitute in 100 ul of distilled H2O for a 1 ug/ul solution.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.