Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYH7B Rabbit anti-Human, Mouse, Rat, Clone: 7W8J3, Novus Biologicals™
SDP

Catalog No. NB168081 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168081 100 μg
NB168082 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168081 Supplier Novus Biologicals Supplier No. NBP316495100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MYH7B Monoclonal antibody specifically detects MYH7B in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen MYH7B
Applications Western Blot
Classification Monoclonal
Clone 7W8J3
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Antigen MLAA-21, MHC14, MYH14, Myosin cardiac muscle beta chain, myosin heavy chain 7B, Myosin heavy chain 7B, cardiac muscle beta isoform, Myosin-7B, Slow A MYH14
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1750-1983 of human MYH7B (A7E2Y1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57644
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.