Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYH8 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB343403 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343403 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343403 Supplier Novus Biologicals Supplier No. NBP241309MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MYH8 Polyclonal antibody specifically detects MYH8 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen MYH8
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Fetal-Myosin Heavy Chain, gtMHC-F, MyHC-peri, myHC-perinatal, MyHC-pn, Myosin Heavy Chain 8, Myosin Heavy Chain, Skeletal Muscle, Perinatal, Myosin, Heavy Chain 8, Skeletal Muscle, Perinatal, Myosin, Heavy Polypeptide 8, Skeletal Muscle, Perinatal, myosin-8
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid peptide near the amino terminus of human MYH8. The immunogen is located within amino acids 60 - 110 of MYH8. Amino Acid Squence: VREDQVFPMNPPKYDKIED
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4626
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.