Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYL4 Antibody (4E8), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB309958 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309958 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309958 Supplier Novus Biologicals Supplier No. NBP242646MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MYL4 Monoclonal antibody specifically detects MYL4 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen MYL4
Applications Western Blot, ELISA
Classification Monoclonal
Clone 4E8
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias ALC1, AMLC, GT1, MLC1, Myosin light chain 1, embryonic muscle/atrial isoform, myosin light chain 4, Myosin light chain alkali GT-1 isoform, myosin, atrial/fetal muscle, light chain, myosin, light chain 4, alkali; atrial, embryonic, myosin, light polypeptide 4, alkali; atrial, embryonic, PRO1957
Host Species Mouse
Immunogen Recombinant human MYL4 (1-197aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 4635
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.