Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYL4 Antibody (OTI1H6), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N272856J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272856J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272856J646 Supplier Novus Biologicals Supplier No. NBP272856JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MYL4 Monoclonal specifically detects MYL4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen MYL4
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1H6
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ALC1, AMLC, GT1, MLC1, Myosin light chain 1, embryonic muscle/atrial isoform, myosin light chain 4, Myosin light chain alkali GT-1 isoform, myosin, atrial/fetal muscle, light chain, myosin, light chain 4, alkali; atrial, embryonic, myosin, light polypeptide 4, alkali; atrial, embryonic, PRO1957
Gene Symbols MYL4
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 2-197 of human MYL4 (NP_002467) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 4635.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.