Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYL7 Antibody (OTI4E7), PE, Novus Biologicals™
SDP

Catalog No. NBP272859PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272859PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272859PE Supplier Novus Biologicals Supplier No. NBP272859PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MYL7 Monoclonal specifically detects MYL7 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen MYL7
Applications Flow Cytometry
Classification Monoclonal
Clone OTI4E7
Conjugate PE
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias atrial isoform, light polypeptide 7, regulatory, MLC-2a, MYL2A, MYLC2AMLC2a, myosin, light chain 7, regulatory
Gene Symbols MYL7
Host Species Mouse
Immunogen Full length recombinant protein of human MYL7 (NP_067046) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 58498.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.