Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYL7 Mouse anti-Human, Clone: OTI4E7, Novus Biologicals™
SDP

Catalog No. NB029848 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB029848 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB029848 Supplier Novus Biologicals Supplier No. NBP203891100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MYL7 Monoclonal specifically detects MYL7 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MYL7
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI4E7
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150
Formulation PBS (pH 7.3), 1.0% BSA and 50% Glycerol
Gene Alias atrial isoform, light polypeptide 7, regulatory, MLC-2a, MYL2A, MYLC2AMLC2a, myosin, light chain 7, regulatory
Host Species Mouse
Immunogen Full length human recombinant protein of human MYL7 (NP_067046) produced in E.coli.
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58498
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.