Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYL9 Antibody (6G11), Novus Biologicals™
SDP

Human Monoclonal Antibody

Supplier:  Novus Biologicals NBP326123100UL

 View more versions of this product

Catalog No. NB298731


Only null left
Add to cart

Description

Description

MYL9 Monoclonal antibody specifically detects MYL9 in Human samples. It is validated for ELISA
Specifications

Specifications

MYL9
Monoclonal
Unconjugated
50% Glycerol, 0.01M PBS, PH 7.4
Human
Affinity purified
RUO
Primary
Human
Purified
ELISA
6G11
ELISA
LC2020 kDa myosin light chain, MLC2Myosin RLC, MRLC1MLC-2C, myosin regulatory light chain 1, Myosin regulatory light chain 2, smooth muscle isoform, Myosin regulatory light chain 9, Myosin regulatory light chain MRLC1, myosin regulatory light polypeptide 9, myosin, light chain 9, regulatory, myosin, light polypeptide 9, regulatory, MYRL2MGC3505
Recombinant Human MYL9 protein [UniProt P24844]
100 μL
Cell Biology, Cytoskeleton Markers, Signal Transduction
10398
Store at -20 to -70C. Avoid freeze-thaw cycles.
IgG2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.