Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MYL9 Antibody (6G11), Novus Biologicals™

Human Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | MYL9 |
---|---|
Clone | 6G11 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
MYL9 Monoclonal antibody specifically detects MYL9 in Human samples. It is validated for ELISASpecifications
MYL9 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
LC2020 kDa myosin light chain, MLC2Myosin RLC, MRLC1MLC-2C, myosin regulatory light chain 1, Myosin regulatory light chain 2, smooth muscle isoform, Myosin regulatory light chain 9, Myosin regulatory light chain MRLC1, myosin regulatory light polypeptide 9, myosin, light chain 9, regulatory, myosin, light polypeptide 9, regulatory, MYRL2MGC3505 | |
Recombinant Human MYL9 protein [UniProt P24844] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
6G11 | |
ELISA | |
Unconjugated | |
Human | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
50% Glycerol, 0.01M PBS, PH 7.4 | |
10398 | |
IgG2 | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title