Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYLIP Antibody (OTI3A2), HRP, Novus Biologicals™
SDP

Catalog No. NBP272862H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272862H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272862H Supplier Novus Biologicals Supplier No. NBP272862H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MYLIP Monoclonal specifically detects MYLIP in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen MYLIP
Applications Western Blot
Classification Monoclonal
Clone OTI3A2
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias band 4.1 superfamily member BZF1, BZF1, cellular modulator of immune recognition (c-MIR), EC 6.3.2.-, Idol, IDOLmyosin regulatory light chain-interacting protein, Inducible degrader of the LDL-receptor, MIRE3 ubiquitin-protein ligase MYLIP, myosin regulatory light chain interacting proteinE3 ubiquitin ligase-inducible degrader of the low density lipoprotein receptor
Gene Symbols MYLIP
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 127-385 of human MYLIP (NP_037394) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 29116.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.