Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYLIP Antibody (OTI3A2), PerCP, Novus Biologicals™
SDP

Catalog No. NB287744 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287744 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287744 Supplier Novus Biologicals Supplier No. NBP272862PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MYLIP Monoclonal antibody specifically detects MYLIP in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen MYLIP
Applications Western Blot
Classification Monoclonal
Clone OTI3A2
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias band 4.1 superfamily member BZF1, BZF1, cellular modulator of immune recognition (c-MIR), EC 6.3.2.-, Idol, IDOLmyosin regulatory light chain-interacting protein, Inducible degrader of the LDL-receptor, MIRE3 ubiquitin-protein ligase MYLIP, myosin regulatory light chain interacting proteinE3 ubiquitin ligase-inducible degrader of the low density lipoprotein receptor
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 127-385 of human MYLIP(NP_037394) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 29116
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.