Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MyoD Antibody (5.8A) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB013936 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013936 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013936 Supplier Novus Biologicals Supplier No. NBP280863

Mouse Monoclonal Antibody

MyoD Monoclonal specifically detects MyoD in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Knockout Validated, Knockdown Validated.

Specifications

Antigen MyoD
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Immunoassay, KnockDown
Classification Monoclonal
Clone 5.8A
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 5 ug/mL, Immunoprecipitation 1 ug/mL, Immunohistochemistry-Frozen 1:10 - 1:500, Knockout Validated, KnockDown Validated
Formulation PBS
Gene Alias bHLHc1BHLHC1, Class C basic helix-loop-helix protein 1, MYF3Myf-3, MYODmyoblast determination protein 1, myogenic differentiation 1, Myogenic factor 3myf-3, PUM
Gene Symbols MYOD1
Host Species Mouse
Immunogen The 5.8A antibody was made against recombinant mouse MyoD protein but it also recognizes human (myf3), rat, and cat homologs. The epitope of this antibody was mapped to a region within aa 180-189 of mouse MyoD (NP_002469).
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Growth and Development, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 4654
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.