Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MyoD Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB338224 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338224 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338224 Supplier Novus Biologicals Supplier No. NBP154153MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MyoD Polyclonal antibody specifically detects MyoD in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen MyoD
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias bHLHc1BHLHC1, Class C basic helix-loop-helix protein 1, MYF3Myf-3, MYODmyoblast determination protein 1, myogenic differentiation 1, Myogenic factor 3myf-3, PUM
Host Species Rabbit
Immunogen A partial recombinant protein made to an N-terminal region of the human MyoD1 protein (within residues 1-150). [Swiss-Prot# P15172]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Growth and Development, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 4654
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.