Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myofibroblast Marker Antibody (PR 2D3), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB378766 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB378766 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB378766 Supplier Novus Biologicals Supplier No. NBP308768AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Myofibroblast Marker Monoclonal antibody specifically detects Myofibroblast Marker in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)

Specifications

Antigen Myofibroblast Marker
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PR 2D3
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Crude homogenate of normal human colorectal mucosa
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.