Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myofibroblast Marker Antibody (PR 2D3), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB309291 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309291 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309291 Supplier Novus Biologicals Supplier No. NBP250524MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Myofibroblast Marker Monoclonal antibody specifically detects Myofibroblast Marker in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation

Specifications

Antigen Myofibroblast Marker
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone PR 2D3
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Crude homogenate of normal colorectal mucosa.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.