Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myogenin Antibody (5FD) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10056510 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056510 0.1 mg
NB10056510T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056510 Supplier Novus Biologicals Supplier No. NB10056510

Mouse Monoclonal Antibody has been used in 9 publications

Myogenin Monoclonal specifically detects Myogenin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Myogenin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 5FD
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunohistochemistry 1:20 - 1:1000, Immunocytochemistry/Immunofluorescence 5 ug/mL, Immunohistochemistry-Paraffin 20 - 40 ug/mL, Immunohistochemistry-Frozen 10 - 20 ug/mL
Gene Accession No. P15173
Gene Alias BHLHC3, bHLHc3Myogenic factor-4; myogenin, Class C basic helix-loop-helix protein 3, myf-4, MYF4myogenin, Myogenic factor 4, MYOGENIN, myogenin (myogenic factor 4)
Gene Symbols MYOG
Host Species Mouse
Immunogen A recombinant protein corresponding to amino acids 30-224 of rat myogenin was used as immunogen (NP_002470).
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Stem Cell Markers, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4656
Test Specificity The 5FD clone has been highly published; users are encouraged to consult the scientific literature for additional application information. In Rh-30, a 34 kDa band should be observed. The can be used for gel shift assays.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.