Learn More
Description
Specifications
Specifications
| Antigen | Myogenin |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 5FD |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 ug/mL, Immunohistochemistry 1:20 - 1:1000, Immunocytochemistry/Immunofluorescence 5 ug/mL, Immunohistochemistry-Paraffin 20 - 40 ug/mL, Immunohistochemistry-Frozen 10 - 20 ug/mL |
| Gene Accession No. | P15173 |
| Gene Alias | BHLHC3, bHLHc3Myogenic factor-4; myogenin, Class C basic helix-loop-helix protein 3, myf-4, MYF4myogenin, Myogenic factor 4, MYOGENIN, myogenin (myogenic factor 4) |
| Gene Symbols | MYOG |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
