Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myogenin Antibody (671038), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB375960 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB375960 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB375960 Supplier Novus Biologicals Supplier No. FAB6686V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Myogenin Monoclonal antibody specifically detects Myogenin in Human, Mouse samples. It is validated for Immunocytochemistry

Specifications

Antigen Myogenin
Applications Immunocytochemistry
Classification Monoclonal
Clone 671038
Conjugate Alexa Fluor 405
Dilution Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias BHLHC3, bHLHc3Myogenic factor-4; myogenin, Class C basic helix-loop-helix protein 3, myf-4, MYF4myogenin, Myogenic factor 4, MYOGENIN, myogenin (myogenic factor 4)
Host Species Mouse
Immunogen E. coli-derived recombinant human Myogenin, Ser124-Asn224, Accession # P15173
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Stem Cell Markers, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4656
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.