Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myogenin Antibody (rMYOG/6297), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB178122 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB178122 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB178122 Supplier Novus Biologicals Supplier No. NBP314012JF549
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Myogenin Monoclonal antibody specifically detects Myogenin in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Myogenin
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rMYOG/6297
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias BHLHC3, bHLHc3Myogenic factor-4; myogenin, Class C basic helix-loop-helix protein 3, myf-4, MYF4myogenin, Myogenic factor 4, MYOGENIN, myogenin (myogenic factor 4)
Host Species Mouse
Immunogen Recombinant full-length human Myogenin protein (Uniprot: P15173)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Stem Cell Markers, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4656
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.