Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myoglobin Antibody (5A2G8) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB11089456 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11089456 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11089456 Supplier Novus Biologicals Supplier No. NB11089456
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Myoglobin Monoclonal specifically detects Myoglobin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Myoglobin
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 5A2G8
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:1000
Formulation PBS with 0.03% Sodium Azide
Gene Alias MGC13548, myoglobin, PVALB
Gene Symbols MB
Host Species Mouse
Immunogen Ni-NTA purified truncated recombinant Myoglobin expressed in E. Coli strain BL21 (DE3)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 4151
Test Specificity Myoglobin (4D7H3)
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.