Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myoneurin Antibody (OTI3B9), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N272864PEC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272864PEC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272864PEC5 Supplier Novus Biologicals Supplier No. NBP272864PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Myoneurin Monoclonal specifically detects Myoneurin in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Myoneurin
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3B9
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias myoneurin, OSZF, ZBTB31SBBIZ1, Zinc finger and BTB domain-containing protein 31, zinc finger protein with BTB/POZ domain, ZNF902
Gene Symbols MYNN
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 10-300 of human Myoneurin (NP_061127) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 55892.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.