Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myosin 7 (MYH7) Rat anti-Mouse, Clone: 4B51E8, MilliporeSigma™
Rat Monoclonal Antibody
$192.35 - $456.56
Specifications
Antigen | Myosin 7 (MYH7) |
---|---|
Clone | 4B51E8 |
Concentration | Please refer to lot specific datasheet. |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABT849-25UG
|
MilliporeSigma
MABT84925UG |
25 μg |
Each for $192.35
|
|
|||||
MABT849-100
|
MilliporeSigma
MABT849100UG |
100 μg |
Each for $456.56
|
|
|||||
Description
Anti-Myosin 7 (MYH7), clone 4B51E8 is a rat monoclonal antibody that detects Mysoin-7 and has been tested for use in Immunofluorescence, Immunohistochemistry (Paraffin), and Western Blotting.Specifications
Myosin 7 (MYH7) | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
KLH-conjugated linear peptide corresponding to 39 amino acids from the C-terminal region of murine Mysoin-7. | |
Primary | |
Stable for 1 year at 2-8°C from the date of receipt. |
4B51E8 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rat | |
Cell Structure | |
Myh7 | |
IgG2a κ | |
Protein G Purified | |
Clone 4B51E8 is a rat monoclonal antibody that detects murine Myosin-7. It targets an epitope within 39 amino acids from the C-terminall half. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title