Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin Heavy Chain 1 Antibody (MY32), Novus Biologicals™
SDP

Catalog No. NBP122811 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
6 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP122811 6 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP122811 Supplier Novus Biologicals Supplier No. NBP122811
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Myosin Heavy Chain 1 Monoclonal antibody specifically detects Myosin Heavy Chain 1 in Human, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen Myosin Heavy Chain 1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MY32
Conjugate Unconjugated
Dilution Simple Western, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin
Formulation Ascites with 0.05% Sodium Azide
Gene Alias DFNA4, FLJ13881, FLJ43092, KIAA2034DKFZp667A1311, MHC16, MYH14 variant protein, MYH17, myosin, Myosin heavy chain 14, Myosin heavy chain, non-muscle IIc, myosin, heavy chain 14, myosin, heavy chain 14, non-muscle, myosin, heavy polypeptide 14, myosin-14, NMHC II-C, NMHC-II-C, nonmuscle myosin heavy chain II-C, Non-muscle myosin heavy chain IIc
Gene Symbols MYH1
Host Species Mouse
Immunogen Rabbit muscle myosin.
Purification Method Ascites
Quantity 6 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4619
Test Specificity This antibody recognizes the heavy chain of human skeletal myosin. It does not stain cardiac or smooth muscle myosin. Rather, it stains the fast twitch (type II) isomyosin molecule.
Target Species Human, Rabbit, Rat, Bovine, Chicken, Guinea Pig, Rabbit
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.