Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin Heavy Chain 1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP215029 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP215029 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP215029 Supplier Novus Biologicals Supplier No. NBP215029
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Myosin Heavy Chain 1 Polyclonal specifically detects Myosin Heavy Chain 1 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen Myosin Heavy Chain 1
Applications Western Blot
Classification Polyclonal
Concentration 0.92mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7), 20% Glycerol, 1% BSA with 0.025% Proclin 300
Gene Accession No. P12882
Gene Alias MyHC-2x, MYH1, MYH1 myosin, heavy chain 1, skeletal muscle, adult MYHa, MYHC 1, MYHC1, MYHC-1, MyHC-2X/D, MYHSA1
Gene Symbols MYH1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human MYH1. The exact sequence is proprietary.
Molecular Weight of Antigen 223 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4619
Test Specificity Mouse. Predicted cross-reactivity based sequence identity: Rhesus Monkey (100%).
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.