Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myosin heavy chain 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$191.80 - $371.50
Specifications
Antigen | Myosin heavy chain 1 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEPA5145318
|
Invitrogen™
PA5145318 |
20 μL |
Each for $191.80
|
|
|||||
FEPA5145317
|
Invitrogen™
PA5145317 |
200 μL |
Each for $371.50
|
|
|||||
Description
Myosin is a protein which is known to interact with actin in muscle and non-muscle cells. It contains two identical heavy chains and four light chains. Myosin molecules consist of two major regions: tails (rods) and heads. They aggregate into filaments through the tail region and interact with actin and with ATP through the head region. Multiple forms of myosin heavy chains exist for each muscle type-skeletal, cardiac, smooth and non-muscle isomyosin forms exist in different types of skeletal muscle, depending on the physiological function of the muscle. They are designated at type I (slow twitch) and type II (fast twitch).Specifications
Myosin heavy chain 1 | |
Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat, Pig | |
P12882, Q5SX40, Q9TV61 | |
100125538, 17879, 287408, 4619 | |
Recombinant Human MYH1 protein, Tyr585-Ala794 (Accession #P12882). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
A530084A17Rik; epididymis luminal protein 71; HEL71; IId; IId/x; MdMs; MHC2X/D; MHC-2X/D; MYH1; MYHa; MYHC; myHC-2x; MyHC-2X/D; MyHC-IId/x; MYHC-IIX; MyHC-IIx/d; MYHSA1; Myhs-f; Myhsf2; Myhs-f2; myosin heavy chain 1; Myosin heavy chain 2x; myosin heavy chain IIx/d; Myosin heavy chain, skeletal muscle, adult 1; myosin, heavy chain 1, skeletal muscle, adult; myosin, heavy polypeptide 1, skeletal muscle, adult; myosin-1; type 2X myosin heavy chain | |
Myh1 | |
IgG | |
Protein A | |
Myh1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title