Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin heavy chain 11 Antibody (160), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB308261 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308261 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308261 Supplier Novus Biologicals Supplier No. NBP289243MFV500
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Myosin heavy chain 11 Monoclonal antibody specifically detects Myosin heavy chain 11 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Myosin heavy chain 11
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 160
Conjugate mFluor Violet 500 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias AAT4, FAA4, MYH11, MYH11 myosin, heavy chain 11, smooth muscle, SMHC, SMMHC, Smooth muscle myosin heavy chain
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the center region of the Human Myosin heavy chain 11.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Hypoxia, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4629
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.