Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin heavy chain 11 Antibody (MYH11/2303R), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB380307 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB380307 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB380307 Supplier Novus Biologicals Supplier No. NBP308486R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Myosin heavy chain 11 Monoclonal antibody specifically detects Myosin heavy chain 11 in Human, Rat, Porcine, Bovine, Canine, Chicken, Feline, Guinea Pig, Rabbit samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Myosin heavy chain 11
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MYH11/2303R
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias AAT4, FAA4, MYH11, MYH11 myosin, heavy chain 11, smooth muscle, SMHC, SMMHC, Smooth muscle myosin heavy chain
Host Species Rabbit
Immunogen Recombinant full-length human Myosin heavy chain 11 protein (Uniprot: P35749)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Hypoxia, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4629
Target Species Human, Rat, Pig, Bovine, Canine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.