Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin heavy chain 11 Antibody (MYH11/923), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N24790PE594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N24790PE594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N24790PE594 Supplier Novus Biologicals Supplier No. NBP247900PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Myosin heavy chain 11 Monoclonal specifically detects Myosin heavy chain 11 in Human, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Myosin heavy chain 11
Applications Flow Cytometry
Classification Monoclonal
Clone MYH11/923
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias AAT4, FAA4, MYH11, MYH11 myosin, heavy chain 11, smooth muscle, SMHC, SMMHC, Smooth muscle myosin heavy chain
Gene Symbols MYH11
Host Species Mouse
Immunogen Recombinant full-length human Myosin heavy chain 11 protein (Uniprot: P35749)
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Hypoxia, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4629.0
Test Specificity Smooth muscle myosin heavy chain (SM-MHC) is a cytoplasmic structural protein, which is a major component of the contractile apparatus in smooth muscle cells. Expression of smooth muscle myosin is developmentally regulated, appearing early in smooth muscle development, and is specific for smooth muscle development. Two isoforms of smooth muscle myosin heavy chain have been identified, designated MHC-1 and MHC-2. The antibody may be useful for the study of breast tumors as the presence of an intact layer of myoepithelial cells is an important feature, which may distinguish benign breast lesions and carcinoma in situ from invasive tumors.
Target Species Human, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.