Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin heavy chain 2 Antibody (TH81 (BGN/04/4481)), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB10065675V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065675V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065675V Supplier Novus Biologicals Supplier No. NB10065675V

Mouse Monoclonal Antibody

Myosin heavy chain 2 Monoclonal specifically detects Myosin heavy chain 2 in Human, Rat, Rabbit samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Myosin heavy chain 2
Applications ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TH81 (BGN/04/4481)
Conjugate DyLight 405
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias fast 2a myosin heavy chain, MYH2A, MYHas8, MyHC-2A, MyHC-IIa, MYHSA2, myosin heavy chain 2, myosin heavy chain 2a, myosin heavy chain IIa, myosin, heavy chain 2, skeletal muscle, adult, myosin, heavy polypeptide 2, skeletal muscle, adult, myosin-2, skeletal muscle, adult 2, type IIA myosin heavy chain
Gene Symbols MYH2
Host Species Mouse
Immunogen Myosin heavy chain from rabbit muscle
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytoskeleton Markers
Primary or Secondary Primary
Gene ID (Entrez) 4620
Test Specificity NB100-65675 reacts strongly with human cardiac myosin and also recognizes rabbit myosin heavy and light chains.
Target Species Human, Rat, Rabbit
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.