Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin Heavy Chain 3 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB189152 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB189152 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB189152 Supplier Novus Biologicals Supplier No. NBP281968AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Myosin Heavy Chain 3 Polyclonal antibody specifically detects Myosin Heavy Chain 3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Myosin Heavy Chain 3
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias HEMHC, Muscle embryonic myosin heavy chain, MYHC-EMB, MYHSE1, Myosin heavy chain, fast skeletal muscle, embryonic, myosin, heavy chain 3, skeletal muscle, embryonic, myosin, heavy polypeptide 3, skeletal muscle, embryonic, myosin, skeletal, heavy chain, embryonic 1, myosin-3, SMHCEMyosin heavy chain 3
Host Species Rabbit
Immunogen Myosin Heavy Chain 3 antibody was raised against a 19 amino acid peptide near the amino terminus of human Myosin Heavy Chain 3. The immunogen is located within amino acids 30 - 80 of Myosin Heavy Chain 3. Amino Acid Squence: DSKEEYAKGKIKSSQDGK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Dendritic Cell Markers, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4621
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.