Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin-Id Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB167163 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167163 100 μg
NB167164 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167163 Supplier Novus Biologicals Supplier No. NBP316034100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Myosin-Id Polyclonal antibody specifically detects Myosin-Id in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen Myosin-Id
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias KIAA0727myr4, myosin ID, myosin-I gamma, myosin-Id
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 180-380 of human Myosin-Id (NP_001290209.1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 4642
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.