Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myosin-Id Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$148.20 - $378.00
Specifications
Antigen | Myosin-Id |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Myosin-Id Polyclonal antibody specifically detects Myosin-Id in Human, Mouse samples. It is validated for Western BlotSpecifications
Myosin-Id | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
KIAA0727myr4, myosin ID, myosin-I gamma, myosin-Id | |
Recombinant fusion protein containing a sequence corresponding to amino acids 180-380 of human Myosin-Id (NP_001290209.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Vision | |
PBS, 50% glycerol, pH7.3 | |
4642 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title