Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myosin Light Chain 2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 5A1F1, Proteintech

Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 602291IG150UL
Description
Thus gene encodes the regulatory light chain associated with cardiac myosin beta heavy chain. Ca+ triggers the phosphorylation of regulatory light chain that in turn triggers contraction. Mutations in this gene are associated with mid-left ventricular chamber type hypertrophic cardiomyopathy.Specifications
| Myosin Light Chain 2 | |
| Monoclonal | |
| 0.7 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P08733, P10916, P51667 | |
| Myl2 | |
| Myosin Light Chain 2 Fusion Protein Ag1356 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 5A1F1 | |
| Unconjugated | |
| MYL2 | |
| MLC 2, MLC 2v, MLC2, MLC2v, MYL2, Myosin Light Chain 2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 17906, 363925, 396690, 4633 | |
| -20°C | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction