Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myosin Light Chain 2 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 5A1F1, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | Myosin Light Chain 2 |
---|---|
Clone | 5A1F1 |
Concentration | 0.7 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6330
![]() ![]() |
Proteintech Group Inc
602291IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Thus gene encodes the regulatory light chain associated with cardiac myosin beta heavy chain. Ca+ triggers the phosphorylation of regulatory light chain that in turn triggers contraction. Mutations in this gene are associated with mid-left ventricular chamber type hypertrophic cardiomyopathy.Specifications
Myosin Light Chain 2 | |
0.7 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
MLC 2, MLC 2v, MLC2, MLC2v, MYL2, Myosin Light Chain 2 | |
Myl2 | |
IgG1 | |
Protein G | |
Antibody |
5A1F1 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Pig, Rat | |
P08733, P10916, P51667 | |
17906, 363925, 396690, 4633 | |
Myosin Light Chain 2 Fusion Protein Ag1356 | |
Primary | |
-20°C | |
MYL2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title