Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin light chain kinase Antibody (28C5), Novus Biologicals™
SDP

Catalog No. NB298733 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298733 100 μL
NB298732 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298733 Supplier Novus Biologicals Supplier No. NBP326122100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Myosin light chain kinase Monoclonal antibody specifically detects Myosin light chain kinase in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA

Specifications

Antigen Myosin light chain kinase
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 28C5
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias DKFZp686I10125, EC 2.7.11, EC 2.7.11.18, FLJ12216, Kinase-related protein, KRP, light polypeptide kinase, MLCK1, MLCK108, MLCKMLCK210, MSTP083, MYLK1, myosin light chain kinase, smooth muscle, smooth muscle myosin light chain kinase, telokin
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Myosin light chain kinase [UniProt Q15746]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 4638
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.