Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myosin light chain kinase Antibody (28C5), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Myosin light chain kinase |
---|---|
Clone | 28C5 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA |
Applications | Western Blot, Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
Myosin light chain kinase Monoclonal antibody specifically detects Myosin light chain kinase in Human samples. It is validated for Western Blot, Flow Cytometry, ELISASpecifications
Myosin light chain kinase | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
DKFZp686I10125, EC 2.7.11, EC 2.7.11.18, FLJ12216, Kinase-related protein, KRP, light polypeptide kinase, MLCK1, MLCK108, MLCKMLCK210, MSTP083, MYLK1, myosin light chain kinase, smooth muscle, smooth muscle myosin light chain kinase, telokin | |
A synthesized peptide derived from Human Myosin light chain kinase [UniProt Q15746] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
28C5 | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Rabbit | |
Protein Kinase | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
4638 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title