Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin Phosphatase, p Thr850, p Thr853 Antibody, Novus Biologicals™
SDP

Catalog No. NBP21318501 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP21318501 0.1 mg
NBP21318505 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP21318501 Supplier Novus Biologicals Supplier No. NBP2131850.1MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Myosin Phosphatase Polyclonal specifically detects Myosin Phosphatase in Human samples. It is validated for Western Blot.

Specifications

Antigen Myosin Phosphatase
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 μ/mL
Formulation 0.1 M Tris-Glycine (pH 7.4), 0.15 M NaCl and 30% Glycerol with 0.05% sodium azide
Gene Alias MBSmyosin phosphatase, target subunit 1, MGC133042, Myosin phosphatase target subunit 1, Myosin phosphatase-targeting subunit 1, MYPT1protein phosphatase 1 regulatory subunit 12A, protein phosphatase 1, regulatory (inhibitor) subunit 12A, Protein phosphatase myosin-binding subunit
Gene Symbols PPP1R12A
Host Species Rabbit
Immunogen Synthetic peptide corresponding to aa845-855, C-EKRRS[pT]GVSFW of chicken MYPT1 with an N-terminal cysteine added for conjugation purposes (Q90623). Phosphorylation site corresponds to Thr853 in human MYPT1.
Quantity 0.1 mg
Primary or Secondary Primary
Gene ID (Entrez) 4659
Test Specificity Recognizes recombinant human truncated MYPT1 phosphorylated on Thr853, MW approx 80kD.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.