Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Myosin Phosphatase, p Thr850, p Thr853 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$463.00 - $706.50
Specifications
Antigen | Myosin Phosphatase |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 0.5-2 μ/mL |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP21318505
![]() |
Novus Biologicals
NBP2131850.05MG |
0.05 mg |
Each for $463.00
|
|
|||||
NBP21318501
![]() |
Novus Biologicals
NBP2131850.1MG |
0.1 mg |
Each for $706.50
|
|
|||||
Description
Myosin Phosphatase Polyclonal specifically detects Myosin Phosphatase in Human samples. It is validated for Western Blot.Specifications
Myosin Phosphatase | |
Western Blot 0.5-2 μ/mL | |
Polyclonal | |
Rabbit | |
0.1 M Tris-Glycine (pH 7.4), 0.15 M NaCl and 30% Glycerol with 0.05% sodium azide | |
4659 | |
Synthetic peptide corresponding to aa845-855, C-EKRRS[pT]GVSFW of chicken MYPT1 with an N-terminal cysteine added for conjugation purposes (Q90623). Phosphorylation site corresponds to Thr853 in human MYPT1. | |
Primary | |
Recognizes recombinant human truncated MYPT1 phosphorylated on Thr853, MW approx 80kD. |
1 mg/mL | |
Western Blot | |
Unconjugated | |
Human | |
MBSmyosin phosphatase, target subunit 1, MGC133042, Myosin phosphatase target subunit 1, Myosin phosphatase-targeting subunit 1, MYPT1protein phosphatase 1 regulatory subunit 12A, protein phosphatase 1, regulatory (inhibitor) subunit 12A, Protein phosphatase myosin-binding subunit | |
PPP1R12A | |
IgG | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title