Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin VIIa Antibody (31A12), Novus Biologicals™
SDP

Catalog No. NB298734 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298734 50 μL
NB298735 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298734 Supplier Novus Biologicals Supplier No. NBP32612150UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Myosin VIIa Monoclonal antibody specifically detects Myosin VIIa in Human samples. It is validated for Flow Cytometry, ELISA

Specifications

Antigen Myosin VIIa
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 31A12
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:200, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias deafness, autosomal dominant 11, deafness, autosomal recessive 2, DFNA11, DFNB2, myosin VIIA, myosin-VIIa, MYOVIIA, MYU7A, USH1Bsevere))
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Myosin VIIa [UniProt Q13402]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 4647
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.