Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

myosin X Antibody (CL8994), Novus Biologicals™
SDP

Catalog No. NB036515 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036515 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036515 Supplier Novus Biologicals Supplier No. NBP288926
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

myosin X Monoclonal specifically detects myosin X in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen myosin X
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone CL8994
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 2-10 ug/ml
Formulation PBS, pH 7.2, containing 40% glycerol
Gene Alias FLJ10639, FLJ21066, FLJ22268, KIAA0799FLJ43256, MGC131988, myosin X, myosin-X, Unconventional myosin-10
Host Species Mouse
Immunogen This antibody was developed against a synthetic peptide corresponding to amino acids: DDAFKDSPNPSEHGHSDQR
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4651
Test Specificity Specificity of myosin X antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.