Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosin Light Chain 2 Rabbit anti-Human, Mouse, Rat, Zebrafish, Clone: SN67-09, Novus Biologicals™
SDP

Catalog No. NBP267649 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267649 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267649 Supplier Novus Biologicals Supplier No. NBP267649

Rabbit Monoclonal Antibody

Myosin Light Chain 2 Monoclonal specifically detects Myosin Light Chain 2 in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen MyosinLightChain2
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SN67-09
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias cardiac ventricular myosin light chain 2, CMH10myosin light chain 2, DKFZp779C0562, MLC2, MLC-2, MLC-2v, myosin regulatory light chain 2, ventricular/cardiac muscle isoform, myosin, light chain 2, regulatory, cardiac, slow, myosin, light polypeptide 2, regulatory, cardiac, slow, regulatory light chain of myosin, RLC of myosin, slow cardiac myosin regulatory light chain 2
Gene Symbols MYL2
Host Species Rabbit
Immunogen Synthetic peptide within Human Myosin Light Chain 2 aa 117-166/ 166. (SwissProt: P10916 Human; SwissProt: P51667 Mouse; SwissProt: P08733 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 4633
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.