Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myosmine, 98%, may contain up to 2% water

Catalog No. aal11886mc
Click to view available options
Quantity:
100 mg
500 mg

CAS: 532-12-7 | C9H10N2 | 146.193 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 532-12-7
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.193
MDL Number MFCD00052019
InChI Key DPNGWXJMIILTBS-UHFFFAOYSA-N
Synonym myosmine, 3-3,4-dihydro-2h-pyrrol-5-yl pyridine, 3-1-pyrrolin-2-yl pyridine, miosmine, unii-9o0a545w4l, 3-4,5-dihydro-3h-pyrrol-2-yl-pyridine, 2-3-pyridyl-1-pyrroline, 3-2-pyrrolin-2-yl pyridine, pyridine, 3-3,4-dihydro-2h-pyrrol-5-yl, pyridine, 3-1-pyrrolin-2-yl
PubChem CID 442649
ChEBI CHEBI:7051
IUPAC Name 3-(3,4-dihydro-2H-pyrrol-5-yl)pyridine
SMILES C1CC(=NC1)C2=CN=CC=C2

Specifications

Melting Point 42°C to 44°C
Boiling Point 82°C to 83°C (0.5 mmHg)
Quantity 100 mg
Beilstein 119254
Formula Weight 146.19
Percent Purity 98%
Assay may contain up to 2% water
Chemical Name or Material Myosmine
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
RTECSNumber UT7660000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.