Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myrosinase 2 Antibody (6H1), Novus Biologicals™
SDP

Catalog No. NB036742 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036742 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036742 Supplier Novus Biologicals Supplier No. NBP289159
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Myrosinase 2 Monoclonal specifically detects Myrosinase 2 in A. thaliana, Plant samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Myrosinase 2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6H1
Concentration 2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin
Formulation PBS (pH7.4), 0.2% BSA, 50% Glycerol with 0.05% Sodium Azide
Gene Alias AtBGLU37, Beta-glucosidase 37, BGLU37, Sinigrinase 2, TGG2, Thioglucosidase 2
Host Species Mouse
Immunogen Synthetic peptide within thaliana Myrosinase 2 aa 350-400. (SwissProt: Q9C5C2 Human)
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3702
Target Species A. thaliana, Plant
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.