Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYST1 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB291906 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291906 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291906 Supplier Novus Biologicals Supplier No. NB10055252F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MYST1 Polyclonal specifically detects MYST1 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen MYST1
Applications Immunoprecipitation, Western Blot (Negative)
Classification Polyclonal
Conjugate FITC
Dilution Immunoprecipitation, Western Blot (Negative)
Formulation PBS
Gene Alias FLJ14040, hMOFhistone acetyltransferase MYST1, KAT8, MOFEC 2.3.1.48, MOZ, YBF2/SAS3, SAS2 and TIP60 protein 1, MYST histone acetyltransferase 1, MYST-1, ortholog of Drosophila males absent on the first (MOF), probable histone acetyltransferase MYST1
Host Species Rabbit
Immunogen Maps to a region between residue 417 and the C-terminus (residue 467) of human males absent on the first (MYST histone acetyltransferase 1) using the numbering given in entry BAB13924.1 (GeneID 84148).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 84148
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.