Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYST3 Antibody (127A), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP250516G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250516G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250516G Supplier Novus Biologicals Supplier No. NBP250516G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MYST3 Monoclonal specifically detects MYST3 in Human samples. It is validated for Western Blot.

Specifications

Antigen MYST3
Applications Western Blot
Classification Monoclonal
Clone 127A
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.3.1.48, EC 3.1.26.4, histone acetyltransferase MYST3, KAT6A, MGC167033, MOZMonocytic leukemia zinc finger protein, MYST histone acetyltransferase (monocytic leukemia) 3, MYST-3, runt-related transcription factor binding protein 2, Runt-related transcription factor-binding protein 2, RUNXBP2, Zinc finger protein 220, ZNF220MOZ, YBF2/SAS3, SAS2 and TIP60 protein 3
Gene Symbols KAT6A
Host Species Mouse
Immunogen GST-MYST3 (172-321aa) recombinant protein human MYST3
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 7994
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.