Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MYST3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP144049 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144049 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144049 Supplier Novus Biologicals Supplier No. NBP144049
Add to Cart
Add to Cart

Goat Polyclonal Antibody

MYST3 Polyclonal specifically detects MYST3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen MYST3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Immunohistochemistry 3.75 ug/ml, Immunohistochemistry-Paraffin 3.75 ug/ml, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_006757.2
Gene Alias EC 2.3.1.48, EC 3.1.26.4, histone acetyltransferase MYST3, KAT6A, MGC167033, MOZMonocytic leukemia zinc finger protein, MYST histone acetyltransferase (monocytic leukemia) 3, MYST-3, runt-related transcription factor binding protein 2, Runt-related transcription factor-binding protein 2, RUNXBP2, Zinc finger protein 220, ZNF220MOZ, YBF2/SAS3, SAS2 and TIP60 protein 3
Gene Symbols KAT6A
Host Species Goat
Immunogen Peptide with sequence C-EKIKDKEETELD corresponding to internal region according to NP_006757.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 7994
Test Specificity Reported variants NP_001092882.1, NP_006757.2, NP_001092883.1 represent identical protein.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.