Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MZT2A Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB145768 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB145768 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB145768 Supplier Novus Biologicals Supplier No. NBP306012JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MZT2A Polyclonal specifically detects MZT2A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MZT2A
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias FAM128A, Family With Sequence Similarity 128, Member A, Mitotic Spindle Organizing Protein 2A, Mitotic-Spindle Organizing Protein 2A, Mitotic-Spindle Organizing Protein Associated With A Ring Of Gamma-Tubulin 2A, MOZART2A
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MZT2A fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 653784
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.