Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(2-Ethylhexyl)-4-methoxybenzamide, 97%, Thermo Scientific™

Catalog No. p-7039707
Click to view available options
Quantity:
250 mg
1 g

Catalog No. AAH59078MD

Supplier: Thermo Scientific Chemicals H59078MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 560090-29-1
Molecular Formula C16H25NO2
Molecular Weight (g/mol) 263.381
MDL Number MFCD03386079
InChI Key OAGPSBYQMCQNOF-UHFFFAOYSA-N
Synonym n-2-ethylhexyl-4-methoxybenzamide, n-2-ethylhexyl 4-methoxyphenyl carboxamide
PubChem CID 4194504
IUPAC Name N-(2-ethylhexyl)-4-methoxybenzamide
SMILES CCCCC(CC)CNC(=O)C1=CC=C(C=C1)OC

Specifications

Quantity 250 mg
Beilstein 23598861
Formula Weight 296.82
Percent Purity 97%
Chemical Name or Material N-(2-Ethylhexyl)-4-methoxybenzamide
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.